Learn More
Medchemexpress LLC HY-A0125A 50mg Medchemexpress, Alizapride hydrochloride CAS:59338-87-3 Purity:>98%
SDP
Catalog No. 502082327
Encompass_Preferred
Medchemexpress, HY-A0125A 50mg Alizapride hydrochloride CAS:59338-87-3 Alizapride hydrochloride is a dopamine receptor antagonist with prokinetic and antiemetic effects which can also be used in the treatment of nausea and vomiting, including postoperative nausea and vomiting. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2327 Supplier Medchemexpress LLC Supplier No. HYA0125A50MG
Medchemexpress, HY-A0125A 50mg Alizapride hydrochloride CAS:59338-87-3 Alizapride hydrochloride is a dopamine receptor antagonist with prokinetic and antiemetic effects which can also be used in the treatment of nausea and vomiting, including postoperative nausea and vomiting. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.