Learn More
Medchemexpress LLC Alizapride hydrochloride | 59338-87-3 | 99.8% | C16H22ClN5O2 | 1 ML
SDP
Catalog No. 5000432409
Encompass_Preferred
Alizapride is a potent antiemetic that functions as a dopamine receptor antagonist. It is also utilized in the treatment of human digestive disorders.
  • Potent antiemetic properties
  • Functions as a dopamine receptor antagonist
  • Useful in treating human digestive disorders
  • Significantly reduces bound IgG-sensitized erythrocytes with splenic macrophages
Catalog No. 50-004-32409 Supplier Medchemexpress LLC Supplier No. HYA0125A10MM/1ML
Alizapride is a potent antiemetic that functions as a dopamine receptor antagonist. It is also utilized in the treatment of human digestive disorders.
  • Potent antiemetic properties
  • Functions as a dopamine receptor antagonist
  • Useful in treating human digestive disorders
  • Significantly reduces bound IgG-sensitized erythrocytes with splenic macrophages