Learn More
Cayman Chemical Alizapride (hydrochloride), Purity: ≥98%, CAS: 59338-87-3, 50 mg
SDP
Catalog No. 502934289
Encompass_Preferred
A dopamine D2 receptor antagonist (Ki = 66-340 nM in radioligand binding assays); selective for dopamine D2 over α1-, α2-, and β-adrenergic receptors (IC50s = >10 µM for all); reduces dopamine-, apomorphine-, or bromocriptine-induced decreases in gastrointestinal transit in rat at 5 mg/kg
Catalog No. 50-293-4289 Supplier Cayman Chemical Supplier No. 2564550 MG
A dopamine D2 receptor antagonist (Ki = 66-340 nM in radioligand binding assays); selective for dopamine D2 over α1-, α2-, and β-adrenergic receptors (IC50s = >10 µM for all); reduces dopamine-, apomorphine-, or bromocriptine-induced decreases in gastrointestinal transit in rat at 5 mg/kg