Learn More
Apexbio Technology LLC Alizarin 72-48-0 1g
SDP
Catalog No. 502240689
Encompass_Preferred
Alizarin (72-48-0) is a small-molecule inhibitor targeting cell proliferation in tumor cell lines It is designed to inhibit cell growth by interfering with regulatory signaling pathways thereby suppressing tumor cell population expansion Alizarin exerts its biological activity primarily through reduction of ERK pathway phosphorylation and induction of cell cycle arrest in S phase In in vitro studies Alizarin demonstrates antiproliferative activity in prostate cancer breast cancer and bone sarcoma cell lines Based on these pharmacological properties Alizarin holds research potential in oncology for investigating mechanisms of tumor cell growth control and in biomedical research as a calcium-specific staining reagent for evaluating bone mineralization and remodeling
This item is not returnable. View return policy
Catalog No. 50-224-0689 Supplier Apexbio Technology LLC Supplier No. A43081000
Alizarin (72-48-0) is a small-molecule inhibitor targeting cell proliferation in tumor cell lines It is designed to inhibit cell growth by interfering with regulatory signaling pathways thereby suppressing tumor cell population expansion Alizarin exerts its biological activity primarily through reduction of ERK pathway phosphorylation and induction of cell cycle arrest in S phase In in vitro studies Alizarin demonstrates antiproliferative activity in prostate cancer breast cancer and bone sarcoma cell lines Based on these pharmacological properties Alizarin holds research potential in oncology for investigating mechanisms of tumor cell growth control and in biomedical research as a calcium-specific staining reagent for evaluating bone mineralization and remodeling