Learn More
Selleck Chemical LLC Alizarin 100mg 72-48-0 Anthraquinonic
SDP
Catalog No. 508108
Encompass_Preferred
Alizarin (Anthraquinonic) strongly inhibits P450 isoform CYP1A1, CYP1A2 and CYP1B1 with IC50 of 6.2 ?M, 10.0 ?M and 2.7 ?M, respectively; weakly inhibits CYP2A6 and CYP2E1, and does not inhibit CYP2C19, CYP3A4 and CYP3A5. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-810-8 Supplier Selleck Chemical LLC Supplier No. S2526100MG
Alizarin (Anthraquinonic) strongly inhibits P450 isoform CYP1A1, CYP1A2 and CYP1B1 with IC50 of 6.2 ?M, 10.0 ?M and 2.7 ?M, respectively; weakly inhibits CYP2A6 and CYP2E1, and does not inhibit CYP2C19, CYP3A4 and CYP3A5. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.