Learn More
Medchemexpress LLC Alizarin complexone | 3952-78-1 | ≥80.0% | 500 MG
SDP
Catalog No. 5000232951
Encompass_Preferred
Alizarin complexone is a calcium-binding fluorescent dye supplied as an analytical reference standard for research and analytical applications. It binds calcium in mineralized tissues and is used for staining, metal ion detection, and chelation assays.
  • Analytical reference standard suitable for research and analytical use.
  • Calcium-binding fluorescent dye for staining mineralized tissue.
  • Useful for metal ion detection and chelation assays.
  • Supplied as a solid crystalline material; brown-orange crystals, insoluble in water.
Catalog No. 50-002-32951 Supplier Medchemexpress LLC Supplier No. HY121075500MG
Alizarin complexone is a calcium-binding fluorescent dye supplied as an analytical reference standard for research and analytical applications. It binds calcium in mineralized tissues and is used for staining, metal ion detection, and chelation assays.
  • Analytical reference standard suitable for research and analytical use.
  • Calcium-binding fluorescent dye for staining mineralized tissue.
  • Useful for metal ion detection and chelation assays.
  • Supplied as a solid crystalline material; brown-orange crystals, insoluble in water.