Learn More
Medchemexpress LLC Alizarin complexone | 3952-78-1 | 98.0% | 100 MG
SDP
Catalog No. 5000231943
Encompass_Preferred
Alizarin complexone is a calcium-binding fluorescent dye used to stain mineralized bone and trace calcium in research applications. It is supplied as a solid powder and as a DMSO solution for analytical and laboratory use.
  • Calcium-binding fluorescent dye for staining mineralized tissue.
  • Useful for bone labeling and calcium tracing in research.
  • Molecular weight 385.32 g/mol; CAS 3952-78-1.
  • Typical purity ~98.0% and soluble in DMSO.
  • Available as solid powder and as 10 mM solution in DMSO.
Catalog No. 50-002-31943 Supplier Medchemexpress LLC Supplier No. HY121075100MG
Alizarin complexone is a calcium-binding fluorescent dye used to stain mineralized bone and trace calcium in research applications. It is supplied as a solid powder and as a DMSO solution for analytical and laboratory use.
  • Calcium-binding fluorescent dye for staining mineralized tissue.
  • Useful for bone labeling and calcium tracing in research.
  • Molecular weight 385.32 g/mol; CAS 3952-78-1.
  • Typical purity ~98.0% and soluble in DMSO.
  • Available as solid powder and as 10 mM solution in DMSO.