Learn More
Medchemexpress LLC Alizarin complexone | 3952-78-1 | 98.0% | 5 G
SDP
Catalog No. 5000233968
Encompass_Preferred
Alizarin complexone is a calcium-binding fluorescent dye provided as an analytical/reference standard for research and analytical applications. It binds calcium crystals to stain mineralized tissue and is used in chromatography and mass spectrometry methods as a reference compound.
  • Calcium-binding fluorescent dye for staining mineralized tissue.
  • Suitable as an analytical reference standard for HPLC, GC, and MS.
  • Used as a chelating agent for metal ion detection.
  • High-purity grades commonly available from suppliers for research use.
  • Supplied as a stable red to brown powder for laboratory handling.
Catalog No. 50-002-33968 Supplier Medchemexpress LLC Supplier No. HY1210755G
Alizarin complexone is a calcium-binding fluorescent dye provided as an analytical/reference standard for research and analytical applications. It binds calcium crystals to stain mineralized tissue and is used in chromatography and mass spectrometry methods as a reference compound.
  • Calcium-binding fluorescent dye for staining mineralized tissue.
  • Suitable as an analytical reference standard for HPLC, GC, and MS.
  • Used as a chelating agent for metal ion detection.
  • High-purity grades commonly available from suppliers for research use.
  • Supplied as a stable red to brown powder for laboratory handling.