Learn More
Medchemexpress LLC Alizarin cyanin green F | 4403-90-1 | 82.8% | 622.58 | 50 MG
SDP
Catalog No. 5000428176
Encompass_Preferred
Alizarin cyanin green F is an alizarin dye that reacts with calcium possibly via sulfonic acid and/or OH groups.
  • Fluorescent dye
  • Histological analysis
  • Histochemistry
Catalog No. 50-004-28176 Supplier Medchemexpress LLC Supplier No. HYD117550MG
Alizarin cyanin green F is an alizarin dye that reacts with calcium possibly via sulfonic acid and/or OH groups.
  • Fluorescent dye
  • Histological analysis
  • Histochemistry