Learn More
Medchemexpress LLC Alizarin red S | 130-22-3 | 95.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12060150MG
Encompass_Preferred
Alizarin Red S sodium is a water-soluble anthraquinone dye used to stain and detect calcium deposits in biological samples and cell culture. It forms colored complexes with calcium ions and is commonly applied in histology and osteogenic differentiation assays.
  • Used to stain calcium deposits and evaluate osteogenic differentiation
  • Water-soluble sodium salt that forms complexes with calcium ions
  • Suitable for histology and cell-culture staining protocols
  • Chemical formula C14H7NaO7S; molecular weight 342.26 g/mol
Catalog No. 50-002-31789