Learn More
Bioworld Alizarin Red S, 25 g
Catalog No. 50199185
Encompass_Preferred
Alizarin Red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage. Alizarin Red S has also been used as a histochemical stain for calcium in cells of non-osteogenic lineage and also a colorometric pH indicator in the range of 3.8-5.4.Appearance:Crystalline powderPhysical State:SolidSolubility:Soluble in water (1 mg/mL).Storage:Store at 4° CMelting Point:287-289 °CBoiling Point:430 °C
Catalog No. 50-199-185 Supplier Bioworld Supplier No. 217700041
Alizarin Red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage. Alizarin Red S has also been used as a histochemical stain for calcium in cells of non-osteogenic lineage and also a colorometric pH indicator in the range of 3.8-5.4.Appearance:Crystalline powderPhysical State:SolidSolubility:Soluble in water (1 mg/mL).Storage:Store at 4° CMelting Point:287-289 °CBoiling Point:430 °C