Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Alizarin Red S, Certified, C.I. DcAr-5 25 GR
SDP

Catalog No. 50980170
Encompass_Preferred

(Alizarin Camine, Mordant Red 3)
CAS #130-22-3 C₁₄H₇O₇SNa.H₂O F.W. 360.28
Solubility: 7.69% Water 0.1% Alcohol

Alizarin Red S (Alizarin Camine, Mordant Red 3)is for Staining Bone, nervous tissue in small invertebrates, and for sections of nervous tissue. Use to differentiate bone from cartilage in mammalian embryos; used with Toluidine Blue (William, Stain Tech., 16, 23 1941).

Catalog No. 50-980-170 Supplier Electron Microscopy Sciences Supplier No. 10360
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Alizarin Camine, Mordant Red 3)
CAS #130-22-3 C₁₄H₇O₇SNa.H₂O F.W. 360.28
Solubility: 7.69% Water 0.1% Alcohol

Alizarin Red S (Alizarin Camine, Mordant Red 3)is for Staining Bone, nervous tissue in small invertebrates, and for sections of nervous tissue. Use to differentiate bone from cartilage in mammalian embryos; used with Toluidine Blue (William, Stain Tech., 16, 23 1941).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.