Learn More
Abcam Alizarin red S, histochemical stain, 25G
Catalog No. 502704675
Encompass_Preferred
MW 360.3 g/mol. Anthraquinone dye used to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage. Has also been used as a histochemical stain for calcium in cells of non-osteogenic lineage and as a colorimetric pH indicator.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4675 Supplier Abcam Supplier No. AB14637425G
MW 360.3 g/mol. Anthraquinone dye used to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage. Has also been used as a histochemical stain for calcium in cells of non-osteogenic lineage and as a colorimetric pH indicator.
The product is subject to the following: Abcam Restricted Use Statement