Learn More
Medchemexpress LLC Alizarin Red S sodium | 130-22-3 | 25 MG
SDP
Catalog No. 5000232757
Encompass_Preferred
Alizarin Red S sodium is an anthraquinone-derived dye that binds calcium ions and is used to stain mineralized deposits in biological samples. It is commonly used in histology and cell biology to assess osteoblast differentiation and bone formation.
  • Stains calcium deposits for histology and cell-based assays.
  • Forms complexes with calcium for visualization of mineralized matrix.
  • Water-soluble sodium salt suitable for aqueous staining procedures.
  • Supplied in small laboratory quantities for research use.
Catalog No. 50-002-32757 Supplier Medchemexpress LLC Supplier No. HY12060125MG
Alizarin Red S sodium is an anthraquinone-derived dye that binds calcium ions and is used to stain mineralized deposits in biological samples. It is commonly used in histology and cell biology to assess osteoblast differentiation and bone formation.
  • Stains calcium deposits for histology and cell-based assays.
  • Forms complexes with calcium for visualization of mineralized matrix.
  • Water-soluble sodium salt suitable for aqueous staining procedures.
  • Supplied in small laboratory quantities for research use.