Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin Red S, Spectrum™ Chemical
SDP

Catalog No. 18600396 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-396 25 g Amber Glass Bottle
18-600-395 10 g Amber Glass Bottle
18-600-397 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-600-396 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AL20025GM
Only null left
Add to Cart
Add to Cart

AL200, 130-22-3, C14H7O7SNa·H2O

Spectrum™ Chemical Alizarin Red S , also known as 1,2-dihydroxyanthraquinone, can be used as a dye for textile fabrics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.25
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
IUPAC Name sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
SMILES [Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O

Specifications

CAS Max % 100%
Grade Reagent
Packaging Amber Glass Bottle
Color Red/Brown to Dark Red
Quantity 25 g
Chemical Name or Material Alizarin Red S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.