Learn More
Alizarin-Red Staining Solution, 100 mL, G-Biosciences

Description
- It forms a complex with calcium ions via chelation, resulting in a red precipitate.
Specifications
Specifications
| CAS | 130-22-3 |
| Color | Red to Rust |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 360.28 |
| Grade | Histological |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.