Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin, Sigma-Aldrich

Catalog No. 01258953 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-953 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-953 Supplier MilliporeSigma Supelco Supplier No. 122777100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alizarin (1, 2-dihydroxyanthraquinone) is an anthraquinone dye and acts as a main chromophore of madder dye. It is used as a dye in the production of lake pigments. Alizarin is a key constituent in extracts of roots of various members of Rubiaceae, Morinda, Gallium, Oldenlandia families.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-48-0
Molecular Weight (g/mol) 240.21 g/mol

Specifications

Color Yellow
pH 5.5 to 6.8, 10.1 to 12.1
Quantity 100 g
Percent Purity 97%
Chemical Name or Material Alizarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.