Learn More
Abcam Alizarin yellow R, Azo dye, 25G
Catalog No. 502704574
Encompass_Preferred
MW 287.23 g/mol. Azo dye used as a pH indicator. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4574 Supplier Abcam Supplier No. AB14654625G
MW 287.23 g/mol. Azo dye used as a pH indicator. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement