Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Alizarin Yellow R, Sodium Salt
SCI_ED

Catalog No. S25133A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25133A 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25133A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 1718-34-9
Molecular Formula C13H8N3NaO5
Linear Formula C13H8N3O5Na
Synonym Mordant Orange 1
InChI Key HKSANXZSKDUDBD-UHFFFAOYSA-M
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+]
IUPAC Name sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Molecular Weight (g/mol) 309.213
PubChem CID 44135758
Formula Weight 635.1811
Packaging Solid
Density >1.0g/mL
Color Brown or Brown/Red to Brown/Yellow
Quantity 10 g
Physical Form Solid
Chemical Name or Material Alizarin Yellow R, Sodium Salt
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.