Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alizarine yellow R | 2243-76-7 | MFCD00007313 | 287.23 | C13H9N3O5 | 250 MG
SDP

Catalog No. 5000227575
Encompass_Preferred

Alizarine Yellow R is an azo dye used primarily as a pH indicator and biological stain. This entry describes the 250 mg research reagent form (CAS 2243-76-7) with molecular formula C13H9N3O5 and molecular weight 287.23.

  • Used as a pH indicator and biological stain.
  • Azo dye with molecular formula C13H9N3O5 and molecular weight 287.23.
  • Supplied as a 250 mg pack suitable for laboratory research.
  • CAS number 2243-76-7 for unambiguous identification.
  • Purity or dye content not specified on the manufacturer's datasheet.

Catalog No. 50-002-27575 Supplier Medchemexpress LLC Supplier No. HY118320250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alizarine Yellow R is an azo dye used primarily as a pH indicator and biological stain. This entry describes the 250 mg research reagent form (CAS 2243-76-7) with molecular formula C13H9N3O5 and molecular weight 287.23.

  • Used as a pH indicator and biological stain.
  • Azo dye with molecular formula C13H9N3O5 and molecular weight 287.23.
  • Supplied as a 250 mg pack suitable for laboratory research.
  • CAS number 2243-76-7 for unambiguous identification.
  • Purity or dye content not specified on the manufacturer's datasheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.