Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alizarine yellow R | 2243-76-7 | MFCD00007313 | 287.23 g/mol | C13H9N3O5 | 1 G
SDP

Catalog No. 5000227688
Encompass_Preferred

Alizarine Yellow R (Mordant orange 1) is an azo dye used as a pH indicator and biological stain for research and analytical applications. It is suitable for detecting pH changes in alkaline ranges and for staining samples in microscopy and analytical workflows. Supplied for laboratory use in small quantities.

  • Functions as a pH indicator in alkaline ranges
  • Useful as a biological stain for microscopy and histology
  • Chemical formula C13H9N3O5 and molecular weight 287.23 g/mol
  • Supplied in small laboratory quantities suitable for research
  • Store at room temperature; keep dry and cool

Catalog No. 50-002-27688 Supplier Medchemexpress LLC Supplier No. HY1183201G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alizarine Yellow R (Mordant orange 1) is an azo dye used as a pH indicator and biological stain for research and analytical applications. It is suitable for detecting pH changes in alkaline ranges and for staining samples in microscopy and analytical workflows. Supplied for laboratory use in small quantities.

  • Functions as a pH indicator in alkaline ranges
  • Useful as a biological stain for microscopy and histology
  • Chemical formula C13H9N3O5 and molecular weight 287.23 g/mol
  • Supplied in small laboratory quantities suitable for research
  • Store at room temperature; keep dry and cool

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.