Learn More
Medchemexpress LLC Alizarine yellow R (mordant orange 1) | 2243-76-7 | MFCD00007313 | 98.0% | 287.23 g/mol | C13H9N3O5 | 500 MG
SDP
Catalog No. 5000227577
Encompass_Preferred
Alizarine Yellow R is a yellow azo dye used primarily as a pH indicator and biological stain. It is supplied as a powdered reagent with a typical purity of 98.0% and a molecular weight of 287.23 g/mol. The product is suitable for laboratory titrations, staining procedures, and dye applications.
  • Azo dye useful as a pH indicator and stain.
  • Typical purity of 98.0% for reliable results.
  • Provided as a 500 mg powder for small-scale use.
  • Molecular weight 287.23 g/mol and formula C13H9N3O5.
Catalog No. 50-002-27577 Supplier Medchemexpress LLC Supplier No. HY118320500MG
Alizarine Yellow R is a yellow azo dye used primarily as a pH indicator and biological stain. It is supplied as a powdered reagent with a typical purity of 98.0% and a molecular weight of 287.23 g/mol. The product is suitable for laboratory titrations, staining procedures, and dye applications.
  • Azo dye useful as a pH indicator and stain.
  • Typical purity of 98.0% for reliable results.
  • Provided as a 500 mg powder for small-scale use.
  • Molecular weight 287.23 g/mol and formula C13H9N3O5.