Learn More
ACROBiosystems ACROBiosystems Human ALK-7 Protein, His Tag
Catalog No. 501956525
Encompass_Preferred
Recombinant Protein;100UG;Human ALK-7, His Tag (AL7-H52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Glu 113 (Accession # Q8NER5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.9 kDa. The protein migrates as 15-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6525 Supplier ACROBiosystems Supplier No. AL7H52H5100UG
Recombinant Protein;100UG;Human ALK-7, His Tag (AL7-H52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Glu 113 (Accession # Q8NER5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.9 kDa. The protein migrates as 15-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.