Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 238MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: ALK/1503. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Anaplastic Large Cell Lymphoma. Cellular Staining: Cytoplasmic and nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human ALK protein fragment (aa1360-1460). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 80kDa (hybrid) ;200kDa (wild type).Chromosome: 2p23. Gene Symbol: ALK. Gene ID: 238. Ensemble: ENSG00000171094. Uniprot: NP_004295.
Catalog No. 50-188-4710