Learn More
Carnabio Usa Inc ALK(C1156Y)/10UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 0853010
Encompass
ALK [C1156Y], Cytoplasmic domain, Mutant [C1156Y] , Amino Acid 1058-1620(end), BAG10812.1, 90 kDa, Insect (sf21)
Catalog No. 50-156-020