Learn More
Medchemexpress LLC Alk degrader (PROTAC) | 2229037-19-6 | 98.0% | 1002.57 | C49H60ClN9O10S | 5MG
SDP
Catalog No. 5000239456
Encompass_Preferred
TL13-112 is a PROTAC small molecule designed to induce degradation of anaplastic lymphoma kinase (ALK) for laboratory research applications. It is supplied as solid material and as a DMSO solution, and is intended for in vitro research use only.
  • Degrades anaplastic lymphoma kinase (ALK) via a targeted PROTAC mechanism.
  • High purity: 98.0% listed by supplier.
  • Molecular weight: 1002.57; formula: C49H60ClN9O10S.
  • Available as solid quantities and as 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-39456 Supplier Medchemexpress LLC Supplier No. HY1239195MG
TL13-112 is a PROTAC small molecule designed to induce degradation of anaplastic lymphoma kinase (ALK) for laboratory research applications. It is supplied as solid material and as a DMSO solution, and is intended for in vitro research use only.
  • Degrades anaplastic lymphoma kinase (ALK) via a targeted PROTAC mechanism.
  • High purity: 98.0% listed by supplier.
  • Molecular weight: 1002.57; formula: C49H60ClN9O10S.
  • Available as solid quantities and as 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.