Learn More
Carnabio Usa Inc ALK(F1174L)/10UG/CYTOPLASMICA
Supplier:  Carnabio Usa Inc 0851910
Encompass
ALK [F1174L], Cytoplasmic domain, Mutant [F1174], Amino Acid 1058-1620, BAG10812.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0678