Learn More
Carnabio Usa Inc ALK(F1174L)/5UG/CYTOPLASMICAL
Supplier:  Carnabio Usa Inc 085195
Encompass
ALK [F1174L], Cytoplasmic domain, Mutant [F1174], Amino Acid 1058-1620, BAG10812.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0326