Learn More
Cayman Chemical ALK InhIbItr 2 10mg
SDP
Catalog No. 502909630
Encompass_Preferred
An inhibitor of TSSK2 (IC50 = 37 nM)
Catalog No. 50-290-9630 Supplier Cayman Chemical Supplier No. 3682310 MG
An inhibitor of TSSK2 (IC50 = 37 nM)