Learn More
Diagnostic Biosystems ALK PHOS ENHANCER Alkaline Phosphatase Enhancer
Catalog No. 502407626
Encompass_Preferred
This product is used as a signal enhancer in conjunction with alkaline phosphatase based immunostaining systems.\n\nAlkaline Phosphatase, an enzyme derived from bovine intestinal mucosa, is often used as a label for in situ hybridization, immunohistochemistry, Southern and Northern hybridization and DNA sequencing. Detection of this enzyme requires the generation of an insoluble colored reaction end product. Stronger the signal, better the staining. DBS introduces a new stable solution to increase the signal generated by alkaline phosphatase several fold. A single treatment of tissue sections with enhancer before the treatment with  substrate/chromogen enables the visualization of difficult to localize antigens. This enhancer works well with Fast Red but can be used with BCIP/NBT and BCIP/INT also. Specimens stained with the above substrates/chromogens can not be dehydrated in ethanol and hence should be mounted in aqueous based mounting medium like C/C Mount (DBS catalog # K002).
Catalog No. 50-240-7626 Supplier Diagnostic Biosystems Supplier No. K011
This product is used as a signal enhancer in conjunction with alkaline phosphatase based immunostaining systems.\n\nAlkaline Phosphatase, an enzyme derived from bovine intestinal mucosa, is often used as a label for in situ hybridization, immunohistochemistry, Southern and Northern hybridization and DNA sequencing. Detection of this enzyme requires the generation of an insoluble colored reaction end product. Stronger the signal, better the staining. DBS introduces a new stable solution to increase the signal generated by alkaline phosphatase several fold. A single treatment of tissue sections with enhancer before the treatment with  substrate/chromogen enables the visualization of difficult to localize antigens. This enhancer works well with Fast Red but can be used with BCIP/NBT and BCIP/INT also. Specimens stained with the above substrates/chromogens can not be dehydrated in ethanol and hence should be mounted in aqueous based mounting medium like C/C Mount (DBS catalog # K002).