Learn More
Lee Biosolutions Inc Alkaline Phosphatase CIP/CIAP, 50 kU, Lyophilized, Highly Purified, Calf Intestines, Dephosphorylation of DNA, RNA, PCR products
Catalog No. NC2868278
Encompass_Preferred
Medix Biochemica USA's Calf Intestinal Alkaline Phosphatase (CIP/CIAP) is a highly purified hydrolase enzyme sourced from calf intestines, providing 50 kU of activity. This enzyme is integral for various molecular biology applications, particularly in DNA and RNA manipulation. It effectively removes phosphate groups from nucleic acids, preventing issues during cloning and improving sequencing accuracy. Its use extends to PCR product cleanup and clinical diagnostics.
  • Sourced from calf intestines
  • Activity: 50 kU
  • Highly purified hydrolase
  • Removes 5' and 3' phosphates
  • Optimal assay temperature: 37°C
  • Optimal assay pH: 10.35
  • Ideal for cloning and dephosphorylation
  • Applicable in DNA sequencing and SNP analysis
  • Lyophilized physical state
  • Compatible with spectral, analytical diagnostics
Catalog No. NC2868278 Supplier Lee Biosolutions Inc Supplier No. 4802050
Medix Biochemica USA's Calf Intestinal Alkaline Phosphatase (CIP/CIAP) is a highly purified hydrolase enzyme sourced from calf intestines, providing 50 kU of activity. This enzyme is integral for various molecular biology applications, particularly in DNA and RNA manipulation. It effectively removes phosphate groups from nucleic acids, preventing issues during cloning and improving sequencing accuracy. Its use extends to PCR product cleanup and clinical diagnostics.
  • Sourced from calf intestines
  • Activity: 50 kU
  • Highly purified hydrolase
  • Removes 5' and 3' phosphates
  • Optimal assay temperature: 37°C
  • Optimal assay pH: 10.35
  • Ideal for cloning and dephosphorylation
  • Applicable in DNA sequencing and SNP analysis
  • Lyophilized physical state
  • Compatible with spectral, analytical diagnostics