Learn More
Sino Biological Recombinant Human ACVR1 Protein (R206H, GST Tag), Active
Catalog No. 502576056
Encompass_Preferred
Recombinant human ALK2 (R206H) (147-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-6056 Supplier Sino Biological Supplier No. A0612BG500
Recombinant human ALK2 (R206H) (147-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.