Learn More
Medchemexpress LLC HY-112815 10mg Medchemexpress, ALK2-IN-2 CAS:2254409-25-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11281510MG
Encompass_Preferred
Medchemexpress, HY-112815 10mg ALK2-IN-2 CAS:2254409-25-9 ALK2-IN-2 is a potent and selective inhibitor of activin receptor-like kinase 2 (ALK2) with an IC50 of 9 nM, and over 700-fold selectivity against ALK3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9180