Learn More
TARGETMOL CHEMICALS INC ALK2-IN-2 5MG
SDP
Catalog No. 502581739
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ALK2-IN-2 is a potent and selective inhibitor of activin receptor-like kinase 2 (ALK2) (IC50 9 nM) inhibiting ALK2 700-fold more than ALK3. purity: 99%
Catalog No. 50-258-1739 Supplier TARGETMOL CHEMICALS INC Supplier No. T102875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ALK2-IN-2 is a potent and selective inhibitor of activin receptor-like kinase 2 (ALK2) (IC50 9 nM) inhibiting ALK2 700-fold more than ALK3. purity: 99%