Learn More
Eurofins Discoverx ALK4, active, 250 µg. (14-614M)
Supplier:  Eurofins Discoverx 14614M
Encompass
N-terminal GST-tagged recombinant, human ALK4 amino acids 150-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9056