Learn More
Cayman Chemical ALK4 EXrAClular DomaInhu 100ug
SDP
Catalog No. 502948641
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged ALK4 expressed in HEK293 cells • Amino acids: 24-126 • MW: 13 kDa
Catalog No. 50-294-8641 Supplier Cayman Chemical Supplier No. 31846100 UG
Source: Active recombinant human C-terminal His-tagged ALK4 expressed in HEK293 cells • Amino acids: 24-126 • MW: 13 kDa