Learn More
Medchemexpress LLC ALK4290 (AKST4290) | 1251528-23-0 | >99.8% | 512.04 | 100 MG
SDP
Catalog No. 5000347962
Encompass_Preferred
ALK4290 (AKST4290) is a potent and orally active CCR3 inhibitor, derived from patent US20130261153A1, compound Example 2, demonstrating a Ki of 3.2 nM for human CCR3. This compound is suitable for research related to neovascular age-related macular degeneration and Parkinsonism.
  • Potent and orally active CCR3 inhibitor.
  • Exhibits a Ki of 3.2 nM for hCCR3.
  • Used in research for neovascular age-related macular degeneration.
  • Used in research for Parkinsonism.
  • Available in high purity (99.79%).
  • Offered in both solid and solution forms for research use.
Catalog No. 50-003-47962 Supplier Medchemexpress LLC Supplier No. HY136788100MG
ALK4290 (AKST4290) is a potent and orally active CCR3 inhibitor, derived from patent US20130261153A1, compound Example 2, demonstrating a Ki of 3.2 nM for human CCR3. This compound is suitable for research related to neovascular age-related macular degeneration and Parkinsonism.
  • Potent and orally active CCR3 inhibitor.
  • Exhibits a Ki of 3.2 nM for hCCR3.
  • Used in research for neovascular age-related macular degeneration.
  • Used in research for Parkinsonism.
  • Available in high purity (99.79%).
  • Offered in both solid and solution forms for research use.