Learn More
Medchemexpress LLC ALK4290 (AKST4290) | 1251528-23-0 | 99.8% | 512.04 | 50 MG
SDP
Catalog No. 5000349119
Encompass_Preferred
ALK4290 (AKST4290) is a potent and orally active CCR3 inhibitor with a Ki of 3.2 nM for hCCR3. It can be used for the research of neovascular age-related macular degeneration and Parkinsonism.
  • Potent and orally active CCR3 inhibitor
  • Ki of 3.2 nM for hCCR3
  • Suitable for research on neovascular age-related macular degeneration
  • Suitable for research on Parkinsonism
  • Has a purity of 99.79%
  • Molecular weight is 512.04
  • Appears as a solid, white to off-white substance
Catalog No. 50-003-49119 Supplier Medchemexpress LLC Supplier No. HY13678850MG
ALK4290 (AKST4290) is a potent and orally active CCR3 inhibitor with a Ki of 3.2 nM for hCCR3. It can be used for the research of neovascular age-related macular degeneration and Parkinsonism.
  • Potent and orally active CCR3 inhibitor
  • Ki of 3.2 nM for hCCR3
  • Suitable for research on neovascular age-related macular degeneration
  • Suitable for research on Parkinsonism
  • Has a purity of 99.79%
  • Molecular weight is 512.04
  • Appears as a solid, white to off-white substance