Learn More
Cayman Chemical ALK5 EXrAClular DomaInhu 100ug
SDP
Catalog No. 502948419
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged ALK5 expressed in HEK293 cells • Amino acids: 34-125 • MW: 38.1 kDa
Catalog No. 50-294-8419 Supplier Cayman Chemical Supplier No. 31847100 UG
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged ALK5 expressed in HEK293 cells • Amino acids: 34-125 • MW: 38.1 kDa