Learn More
Enzo Life Sciences ALK5 Inhibitor II (1mg). CAS: 446859-33-2
SDP
Supplier:  Enzo Life Sciences ALX270445M001
Encompass_Preferred
Cell permeable, potent, selective and ATP-competitive inhibitor of TGF-β RI kinase (IC50=23nM, 4nM and 18nM for binding, auto-phosphorylation and cellular assay in HepG2 cells of TGF-β RI kinase, respectively). Minimally affects a panel of 9 closely related kinases including p38 MAPK (IC50>16µM). Alternative name: TGF-beta RI Kinase Inhibitor II, 2-(3-(6-Methylpyridin-2-yl)-1H-pyrazol-4-yl)-1,5-naphthyridine. Purity: ≥97% (HPLC). Solubility: 5mg/ml soluble in DMSO or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8571