Learn More
Cayman Chemical ALK5 Inhibitor II (hydrochloride), Purity: ≥98%, CAS: 2319939-07-4, 10 mg
SDP
Catalog No. 502971281
Encompass_Preferred
A selective inhibitor of the TGF-β type 1 receptor ALK5 (IC50s = 4, 18, and 23 nM for ALK5 autophosphorylation, TGF-β cellular assay, and ALK5 binding in HepG2 cells, respectively); used to induce stem cell pluripotency by replacing the reprogramming transcription factor Sox2 via inhibition of the TGF-β signaling pathway and induction of Nanog transcription
Catalog No. 50-297-1281 Supplier Cayman Chemical Supplier No. 1552610 MG
A selective inhibitor of the TGF-β type 1 receptor ALK5 (IC50s = 4, 18, and 23 nM for ALK5 autophosphorylation, TGF-β cellular assay, and ALK5 binding in HepG2 cells, respectively); used to induce stem cell pluripotency by replacing the reprogramming transcription factor Sox2 via inhibition of the TGF-β signaling pathway and induction of Nanog transcription