Learn More
Sino Biological Anti-TGF-beta RI Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10459T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human alk5 / TGFBR1 (rh alk5 / TGFBR1; Catalog#10459-H20B; P36897-1; Thr200-Met503). alk5 / TGFBR1 specific IgG was purified by Human alk5 / TGFBR1 affinity chromatography.
Catalog No. 50-224-6321