Learn More
Eurofins Discoverx ALK6, active, 250 µg. (14-941M)
Supplier:  Eurofins Discoverx 14941M
Encompass
N-terminal 6xHis-tagged recombinant, human ALK6 amino acids 148-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9456