Learn More
Cayman Chemical ALK7 EXrAClular DomaInhu 200ug
SDP
Catalog No. 502947522
Encompass_Preferred
Source: Recombinant C-terminal human IgG1 Fc-tagged ALK7 expressed in HEK293 cells • Amino acids: 22-113 • MW: 36.6 kDa
Catalog No. 50-294-7522 Supplier Cayman Chemical Supplier No. 33731200 UG
Source: Recombinant C-terminal human IgG1 Fc-tagged ALK7 expressed in HEK293 cells • Amino acids: 22-113 • MW: 36.6 kDa