Learn More
Alka-M Photometer Tablet Reagent, Lovibond™
Catalog No. 09050122 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
09-050-122 10 mL
1 options
Catalog No. 09-050-122 Supplier Tintometer Group Supplier No. 513211BT
Add to Cart
Add to Cart
Used with Lovibond colorimeters for the measurement of alkalinity-M and acid capacity.
  • High accuracy
  • Blister packaging provides resistance to most climactic conditions and easy, mess-free handling
  • Precise reagent dosing
  • Significantly longer shelf-life than powders and liquids
Specifications
pH Range 0 to 14
Volume (Metric) 10 mL
No. per Pack 250 Pk.
Type Alkalinity Reagent