Description
- High accuracy
- Blister packaging provides resistance to most climactic conditions and easy, mess-free handling
- Precise reagent dosing
- Significantly longer shelf-life than powders and liquids
Specifications
Specifications
| pH Range | 0 to 14 |
| Volume (Metric) | 10 mL |
| No. per Pack | 100 Pk. |
| Type | Alka-P Reagent |