Learn More
Pharmacal Research Laboratories Alka-Det 2, 30 Gal
SDP
Catalog No. NC0744005
Encompass
Alka-Det 2 is a chlorinated cage and bottle washing alkaline detergent. This product is a good choice when dealing with moderate amounts of soil loads.
Catalog No. NC0744005 Supplier Pharmacal Research Laboratories Supplier No. 09230F
Alka-Det 2 is a chlorinated cage and bottle washing alkaline detergent. This product is a good choice when dealing with moderate amounts of soil loads.