Learn More
Inorganic Ventures ICP ALKALI ALKALINE STD 125mL
SDP
Catalog No. 50723177
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Analyte: Al, Ba, Bi, Cs, In, Mg, Rb, Na, As, Be, Ca, Ga, Li, K, Se, and Sr 100 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-177 Supplier Inorganic Ventures Supplier No. CCS4125ML
Matrix: Nitric Acid (HNO3), Analyte: Al, Ba, Bi, Cs, In, Mg, Rb, Na, As, Be, Ca, Ga, Li, K, Se, and Sr 100 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search