Learn More
Medchemexpress LLC Alkali blue 6B monos 100g
SDP
Catalog No. 5000206421
Encompass_Preferred
Alkali blue 6B monosodium (IND) is a basic dye that can be used as a reagent in biochemical and medical research Alkali blue 6B monosodium (IND) interacts with various proteins and can be used in protein adsorption studies Alkali blue 6B monosodium (IND) contains SO3H- NH and OH groups that may react with divalent heavy metal ions and can be used for the removal of heavy metals from aqua[1][2]
Catalog No. 50-002-06421 Supplier Medchemexpress LLC Supplier No. HYW110925100G
Alkali blue 6B monosodium (IND) is a basic dye that can be used as a reagent in biochemical and medical research Alkali blue 6B monosodium (IND) interacts with various proteins and can be used in protein adsorption studies Alkali blue 6B monosodium (IND) contains SO3H- NH and OH groups that may react with divalent heavy metal ions and can be used for the removal of heavy metals from aqua[1][2]