Learn More
Abcam Alkali blue 6B, pH indicator, 25G
Catalog No. 502704772
Encompass_Preferred
MW 595.7 g/mol. pH indicator. Visual transition interval pH 9.4 (Blue) to pH 14.0 (Red). Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4772 Supplier Abcam Supplier No. AB14627025G
MW 595.7 g/mol. pH indicator. Visual transition interval pH 9.4 (Blue) to pH 14.0 (Red). Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement